Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-cyanothiophene 98.0+%, TCI America™
SDP

Catalog No. B42121G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42121G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B42121G Supplier TCI America Supplier No. B42121G
Add to Cart
Add to Cart

Specifications

CAS 18791-99-6
Melting Point 49°C
Color Yellow
Molecular Formula C5H2BrNS
Quantity 1 g
MDL Number MFCD00114951
Synonym 4-Bromothiophene-2-carbonitrile
InChI Key DJYVXBJLHPKUKS-UHFFFAOYSA-N
SMILES C1=C(SC=C1Br)C#N
IUPAC Name 4-bromothiophene-2-carbonitrile
Molecular Weight (g/mol) 188.042
PubChem CID 14292355
Formula Weight 188.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-cyanothiophene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.