Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-fluorobenzyl Chloride 98.0+%, TCI America™
SDP

Catalog No. B28415G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28415G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B28415G Supplier TCI America Supplier No. B28415G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 85510-82-3
Molecular Formula C7H5BrClF
Molecular Weight (g/mol) 223.47
MDL Number MFCD02094036
InChI Key UDKQGFMDBMYVHI-UHFFFAOYSA-N
Synonym 4-bromo-2-fluorobenzyl chloride, 4-bromo-1-chloromethyl-2-fluorobenzene, 4-bromo-2-fluorobenzylchloride, 4-bromo-1-chloromethyl-2-fluoro-benzene, acmc-209q6a, 2-fluoro-4-bromo-chloromethylbenzene
PubChem CID 2773361
IUPAC Name 4-bromo-1-(chloromethyl)-2-fluorobenzene
SMILES FC1=CC(Br)=CC=C1CCl

Specifications

Color Yellow
Quantity 5 g
UN Number 3265
Formula Weight 223.47
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-2-fluorobenzyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.