Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-Bromo-2-hydroxypyridine | 36953-37-4 | 99.6% | C5H4BrNO | 10 MM/1 ML
SDP

Catalog No. 5000434204
Encompass_Preferred

4-Bromo-2-hydroxypyridine is a chemical intermediate used in research to synthesize M1 muscarinic acetylcholine receptor (M1 mAChR) positive allosteric modulators. It is also applied in studies related to Alzheimer's disease and schizophrenia. This product is supplied as a 10 mM solution in 1 mL of DMSO.

  • Can be used for synthesis of M1 muscarinic acetylcholine receptor positive allosteric modulators
  • Suitable for research on Alzheimer's disease and schizophrenia
  • Molecular weight is 174.00
  • Purity of 99.6%
  • Store powder at -20°C for up to 3 years
  • Store in solvent at -80°C for up to 6 months
  • Appearance is white to light brown solid

Catalog No. 50-004-34204 Supplier Medchemexpress LLC Supplier No. HY6503610MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

4-Bromo-2-hydroxypyridine is a chemical intermediate used in research to synthesize M1 muscarinic acetylcholine receptor (M1 mAChR) positive allosteric modulators. It is also applied in studies related to Alzheimer's disease and schizophrenia. This product is supplied as a 10 mM solution in 1 mL of DMSO.

  • Can be used for synthesis of M1 muscarinic acetylcholine receptor positive allosteric modulators
  • Suitable for research on Alzheimer's disease and schizophrenia
  • Molecular weight is 174.00
  • Purity of 99.6%
  • Store powder at -20°C for up to 3 years
  • Store in solvent at -80°C for up to 6 months
  • Appearance is white to light brown solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.