Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-iodoaniline, 97%

Catalog No. AC397910050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC397910050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC397910050 Supplier Thermo Scientific Chemicals Supplier No. 397910050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 66416-72-6 | C6H5BrIN | 297.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66416-72-6
Molecular Formula C6H5BrIN
Molecular Weight (g/mol) 297.91
MDL Number MFCD02093764
InChI Key HHTYEQWCHQEJNV-UHFFFAOYSA-N
Synonym 4-bromo-2-iodo-phenylamine, 4-bromo-2-iodobenzenamine, 2-iodo-4-bromoaniline, benzenamine,4-bromo-2-iodo, benzenamine, 4-bromo-2-iodo, pubchem3784, acmc-1b9hb, 4-bromo-2-iodophenylamine, asischem t54343, 4-bromo-2-iodo-benzenamine
PubChem CID 823934
IUPAC Name 4-bromo-2-iodoaniline
SMILES C1=CC(=C(C=C1Br)I)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 69°C to 72°C
Color Gray to Purple
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H3(I)NH2
Quantity 5 g
Formula Weight 297.91
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 4-Bromo-2-iodoaniline
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye damage.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.