Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-iodophenol 98.0+%, TCI America™
SDP

Catalog No. B49575G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49575G 5 g
B49571G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49575G Supplier TCI America Supplier No. B49575G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 207115-22-8
Molecular Formula C6H4BrIO
Molecular Weight (g/mol) 298.905
MDL Number MFCD09029644
InChI Key UXIULWIJWDJDQD-UHFFFAOYSA-N
PubChem CID 10924532
IUPAC Name 4-bromo-2-iodophenol
SMILES C1=CC(=C(C=C1Br)I)O

Specifications

Melting Point 75°C
Quantity 5 g
Formula Weight 298.91
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-2-iodophenol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.