Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-2-methoxybenzyl alcohol, 97%, Thermo Scientific™

Catalog No. AAH2733906
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2733906 5 g
AAH2733903 1 g
AAH2733914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2733906 Supplier Thermo Scientific Chemicals Supplier No. H2733906

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used in azaquinone-methide-mediated depolymerization of aromatic carbamate oligomers.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17102-63-5
Molecular Formula C8H9BrO2
Molecular Weight (g/mol) 217.06
MDL Number MFCD07369762
InChI Key HBEIHPSICGGZIF-UHFFFAOYSA-N
PubChem CID 15128243
IUPAC Name (4-bromo-2-methoxyphenyl)methanol
SMILES COC1=C(CO)C=CC(Br)=C1

Specifications

Melting Point 66°C to 70°C
Quantity 5 g
Synonym 4-bromo-2-methoxybenzyl alcohol, 4-bromo-2-methoxyphenyl methanol, 4-bromo-2-methoxy-phenyl-methanol, benzenemethanol, 4-bromo-2-methoxy, acmc-209e2a, 2-methoxy-4-bromobenzyl alcohol, benzenemethanol,4-bromo-2-methoxy
Solubility Information Insoluble in water.
Formula Weight 217.06
Percent Purity 97%
Chemical Name or Material 4-Bromo-2-methoxybenzyl alcohol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.