Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (4-bromo-2-(methylamino)phenyl)diethylphosphine oxide | 2685825-06-1 | MFCD35092689 | 99.8% | 290.14 | C11H17BrNOP | 1 G
SDP

Catalog No. 5000281113
Encompass_Preferred

(4-Bromo-2-(methylamino)phenyl)diethylphosphine oxide is an aryl diethylphosphine oxide used as a drug intermediate for the synthesis of various active compounds. It is supplied for research use with batch-specific analytical documentation and safety information to support synthetic and medicinal chemistry workflows.

  • High purity: 99.8% (LCMS) as reported on the batch COA.
  • Chemical formula: C11H17BrNOP.
  • Molecular weight: 290.14 g/mol.
  • Provided with certificate of analysis and safety data sheet.
  • Suitable as a building block in small-molecule synthesis.
  • Available in a 1 G pack size.

Catalog No. 50-002-81113 Supplier Medchemexpress LLC Supplier No. HYW4415921G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(4-Bromo-2-(methylamino)phenyl)diethylphosphine oxide is an aryl diethylphosphine oxide used as a drug intermediate for the synthesis of various active compounds. It is supplied for research use with batch-specific analytical documentation and safety information to support synthetic and medicinal chemistry workflows.

  • High purity: 99.8% (LCMS) as reported on the batch COA.
  • Chemical formula: C11H17BrNOP.
  • Molecular weight: 290.14 g/mol.
  • Provided with certificate of analysis and safety data sheet.
  • Suitable as a building block in small-molecule synthesis.
  • Available in a 1 G pack size.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.