Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (4-bromo-2-(methylamino)phenyl)diethylphosphine oxide | 2685825-06-1 | MFCD35092689 | 99.8% | 290.14 | C11H17BrNOP | 25 G
SDP

Catalog No. 5000281074
Encompass_Preferred

This compound, (4-bromo-2-(methylamino)phenyl)diethylphosphine oxide, is a phosphine oxide intermediate used in medicinal chemistry and organic synthesis for the preparation of active pharmaceutical compounds. It is supplied with batch documentation and intended for laboratory research and drug development use.

  • High purity suitable for synthetic applications (99.8%).
  • Chemical formula C11H17BrNOP.
  • CAS number 2685825-06-1.
  • Molecular weight 290.14 g/mol.
  • Available in laboratory sizes from 100 mg to >100 g.
  • Supplied with batch-specific certificate of analysis.
  • Intended as a building block for drug intermediate synthesis.

Catalog No. 50-002-81074 Supplier Medchemexpress LLC Supplier No. HYW44159225G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This compound, (4-bromo-2-(methylamino)phenyl)diethylphosphine oxide, is a phosphine oxide intermediate used in medicinal chemistry and organic synthesis for the preparation of active pharmaceutical compounds. It is supplied with batch documentation and intended for laboratory research and drug development use.

  • High purity suitable for synthetic applications (99.8%).
  • Chemical formula C11H17BrNOP.
  • CAS number 2685825-06-1.
  • Molecular weight 290.14 g/mol.
  • Available in laboratory sizes from 100 mg to >100 g.
  • Supplied with batch-specific certificate of analysis.
  • Intended as a building block for drug intermediate synthesis.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.