Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (4-bromo-2-(methylamino)phenyl)diethylphosphine oxide | 2685825-06-1 | MFCD35092689 | 99.8% | 290.14 | C11H17BrNOP | 250 MG
SDP

Catalog No. 5000280969
Encompass_Preferred

(4-Bromo-2-(methylamino)phenyl)diethylphosphine oxide is a high-purity phosphine oxide intended for use as a synthetic intermediate in laboratory research. It is supplied as a light brown solid with supporting documentation for quality and safe handling.

  • High purity (99.8% by LCMS).
  • Suitable as a drug intermediate and building block for organic synthesis.
  • Chemical formula C11H17BrNOP; molecular weight 290.14.
  • CAS number 2685825-06-1 for unambiguous identification.
  • Supplied with certificate of analysis and safety data sheet.
  • Storage stability: room temperature for solid; specified frozen conditions for solutions.

Catalog No. 50-002-80969 Supplier Medchemexpress LLC Supplier No. HYW441592250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(4-Bromo-2-(methylamino)phenyl)diethylphosphine oxide is a high-purity phosphine oxide intended for use as a synthetic intermediate in laboratory research. It is supplied as a light brown solid with supporting documentation for quality and safe handling.

  • High purity (99.8% by LCMS).
  • Suitable as a drug intermediate and building block for organic synthesis.
  • Chemical formula C11H17BrNOP; molecular weight 290.14.
  • CAS number 2685825-06-1 for unambiguous identification.
  • Supplied with certificate of analysis and safety data sheet.
  • Storage stability: room temperature for solid; specified frozen conditions for solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.