Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (4-bromo-2-(methylamino)phenyl)diethylphosphine oxide | 2685825-06-1 | MFCD35092689 | 99.8% | 290.14 g/mol | C11H17BrNOP | 100 MG
SDP

Catalog No. 5000281762
Encompass_Preferred

(4-Bromo-2-(methylamino)phenyl)diethylphosphine oxide is a phosphine-oxide drug intermediate used in medicinal chemistry for the synthesis of active small molecules. It is supplied at high reported purity for research and analytical applications and is offered in multiple laboratory package sizes, including a 100 MG vial.

  • High purity (99.83%).
  • Molecular weight 290.14 g/mol.
  • Molecular formula C11H17BrNOP.
  • Available in small laboratory packages including 100 MG.
  • Storage stability data provided for bulk and in-solvent conditions.
  • Intended for research use only.

Catalog No. 50-002-81762 Supplier Medchemexpress LLC Supplier No. HYW441592100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(4-Bromo-2-(methylamino)phenyl)diethylphosphine oxide is a phosphine-oxide drug intermediate used in medicinal chemistry for the synthesis of active small molecules. It is supplied at high reported purity for research and analytical applications and is offered in multiple laboratory package sizes, including a 100 MG vial.

  • High purity (99.83%).
  • Molecular weight 290.14 g/mol.
  • Molecular formula C11H17BrNOP.
  • Available in small laboratory packages including 100 MG.
  • Storage stability data provided for bulk and in-solvent conditions.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.