Learn More
Medchemexpress LLC (4-bromo-2-(methylamino)phenyl)diethylphosphine oxide | 2685825-06-1 | MFCD35092689 | 99.8% | 290.14 g/mol | C11H17BrNOP | 5 G
SDP
Catalog No. 5000281399
Encompass_Preferred
(4-Bromo-2-(methylamino)phenyl)diethylphosphine oxide is a phosphine-oxide small molecule used as a drug intermediate in medicinal chemistry and organic synthesis. Supplied as a solid for research and development, it supports downstream synthesis of biologically active compounds.
  • High purity suitable for synthesis (99.8%).
  • Molecular formula C11H17BrNOP, molecular weight 290.14 g/mol.
  • Solid appearance, light brown to brown.
  • Multiple package sizes available for R&D workflows.
  • Stable at room temperature as a solid; defined storage stability in solvent.
  • Intended for research use and synthetic applications.
Catalog No. 50-002-81399 Supplier Medchemexpress LLC Supplier No. HYW4415925G
(4-Bromo-2-(methylamino)phenyl)diethylphosphine oxide is a phosphine-oxide small molecule used as a drug intermediate in medicinal chemistry and organic synthesis. Supplied as a solid for research and development, it supports downstream synthesis of biologically active compounds.
  • High purity suitable for synthesis (99.8%).
  • Molecular formula C11H17BrNOP, molecular weight 290.14 g/mol.
  • Solid appearance, light brown to brown.
  • Multiple package sizes available for R&D workflows.
  • Stable at room temperature as a solid; defined storage stability in solvent.
  • Intended for research use and synthetic applications.