Learn More
Medchemexpress LLC (4-bromo-2-(methylamino)phenyl)diethylphosphine oxide | 2685825-06-1 | MFCD35092689 | 99.8% | 290.14 g/mol | C11H17BrNOP | 50 G
SDP
Catalog No. 5000281075
Encompass_Preferred
(4-Bromo-2-(methylamino)phenyl)diethylphosphine oxide is a brominated aniline-derived phosphine oxide used primarily as a drug intermediate in the synthesis of biologically active compounds. It is supplied as a solid with high reported purity and characterized by its molecular formula C11H17BrNOP and molecular weight of 290.14 g/mol.
  • High purity appropriate for chemical synthesis and research use.
  • Solid form simplifies handling and storage.
  • Validated analytical data available (certificate of analysis).
  • Stable under recommended storage conditions for long-term use.
  • Suitable as a building block for medicinal chemistry applications.
Catalog No. 50-002-81075 Supplier Medchemexpress LLC Supplier No. HYW44159250G
(4-Bromo-2-(methylamino)phenyl)diethylphosphine oxide is a brominated aniline-derived phosphine oxide used primarily as a drug intermediate in the synthesis of biologically active compounds. It is supplied as a solid with high reported purity and characterized by its molecular formula C11H17BrNOP and molecular weight of 290.14 g/mol.
  • High purity appropriate for chemical synthesis and research use.
  • Solid form simplifies handling and storage.
  • Validated analytical data available (certificate of analysis).
  • Stable under recommended storage conditions for long-term use.
  • Suitable as a building block for medicinal chemistry applications.