Learn More
Medchemexpress LLC 4-Bromo-2-methylbenzoic acid | 68837-59-2 | 215.05 | 1 ML
SDP
Catalog No. 5000445504
Encompass_Preferred
4-Bromo-2-methylbenzoic acid (4-Bromo-o-toluic acid) is an important compound used in the synthesis of other active compounds. This product is for research use only.
  • Used in organic synthesis
  • Can be used to synthesize 4-allyl-2-methylbenzyl chrysanthemate
  • 4-allyl-2-methylbenzyl chrysanthemate has high toxicity
Catalog No. 50-004-45504 Supplier Medchemexpress LLC Supplier No. HYW00486310MM/1ML
4-Bromo-2-methylbenzoic acid (4-Bromo-o-toluic acid) is an important compound used in the synthesis of other active compounds. This product is for research use only.
  • Used in organic synthesis
  • Can be used to synthesize 4-allyl-2-methylbenzyl chrysanthemate
  • 4-allyl-2-methylbenzyl chrysanthemate has high toxicity