Learn More
Medchemexpress LLC 4-Bromo-2-methylbenzoic acid | 68837-59-2 | MFCD00040905 | 215.05 | 5 G
SDP
Catalog No. 5000445506
Encompass_Preferred
4-Bromo-2-methylbenzoic acid (4-Bromo-o-toluic acid) is an important compound used in the synthesis of other active compounds. It can be utilized to synthesize 4-allyl-2-methylbenzyl chrysanthemate, a compound known for its high toxicity.
  • Used in the synthesis of other active compounds
  • Can be utilized to synthesize 4-allyl-2-methylbenzyl chrysanthemate
Catalog No. 50-004-45506 Supplier Medchemexpress LLC Supplier No. HYW0048635G
4-Bromo-2-methylbenzoic acid (4-Bromo-o-toluic acid) is an important compound used in the synthesis of other active compounds. It can be utilized to synthesize 4-allyl-2-methylbenzyl chrysanthemate, a compound known for its high toxicity.
  • Used in the synthesis of other active compounds
  • Can be utilized to synthesize 4-allyl-2-methylbenzyl chrysanthemate