Learn More
Medchemexpress LLC 4-Bromo-2-nitrobenzoic acid | 99277-71-1 | 246.01 | 10 G
SDP
Catalog No. 5000447394
Encompass_Preferred
4-Bromo-2-nitrobenzoic acid is a biochemical reagent and organic compound utilized for life science research. It serves as an intermediate in the synthesis of Lonafarnib, a compound with potential anticancer properties. This product is intended for research use only.
  • Biochemical reagent for life science research
  • Organic compound
  • Intermediate in Lonafarnib synthesis
  • For research use only
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month
Catalog No. 50-004-47394 Supplier Medchemexpress LLC Supplier No. HYW00272710G
4-Bromo-2-nitrobenzoic acid is a biochemical reagent and organic compound utilized for life science research. It serves as an intermediate in the synthesis of Lonafarnib, a compound with potential anticancer properties. This product is intended for research use only.
  • Biochemical reagent for life science research
  • Organic compound
  • Intermediate in Lonafarnib synthesis
  • For research use only
  • Powder storage: -20°C for 3 years, 4°C for 2 years
  • In solvent storage: -80°C for 6 months, -20°C for 1 month