Learn More
Medchemexpress LLC 4-Bromo-2-nitrobenzoic acid | 99277-71-1 | 246.01 | 25 G
SDP
Catalog No. 5000449482
Encompass_Preferred
4-Bromo-2-nitrobenzoic acid is a biochemical reagent that can be utilized as a biological material or organic compound for various life science research applications. It is also an intermediate in the synthesis of Lonafarnib, a potential anticancer agent. This product is intended for research use only.
  • Can be used as a biological material or organic compound
  • Functions as an intermediate in Lonafarnib synthesis
  • Appears as a solid
  • Color ranges from off-white to light brown
  • Suitable for life science related research
Catalog No. 50-004-49482 Supplier Medchemexpress LLC Supplier No. 25GHYW002727
4-Bromo-2-nitrobenzoic acid is a biochemical reagent that can be utilized as a biological material or organic compound for various life science research applications. It is also an intermediate in the synthesis of Lonafarnib, a potential anticancer agent. This product is intended for research use only.
  • Can be used as a biological material or organic compound
  • Functions as an intermediate in Lonafarnib synthesis
  • Appears as a solid
  • Color ranges from off-white to light brown
  • Suitable for life science related research