Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-bromo-2-nitrotoluene | 60956-26-5 | MFCD00041243 | 98.7% | 216.03 g/mol | C7H6BrNO2 | 100 G
SDP

Catalog No. 5000208790
Encompass_Preferred

4-bromo-2-nitrotoluene is an organic reagent used as an intermediate in organic synthesis and for research applications. It is commonly used as a building block in the synthesis of substituted benzaldehydes, benzyl derivatives, and other nitroaromatic intermediates.

  • used as an intermediate in organic synthesis
  • chemical formula: C7H6BrNO2
  • molecular weight: 216.03 g/mol
  • cas number: 60956-26-5
  • purity typically around 98.7%
  • available in solid form, commonly supplied as a 100 g package

Catalog No. 50-002-08790 Supplier Medchemexpress LLC Supplier No. HYW041357100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-bromo-2-nitrotoluene is an organic reagent used as an intermediate in organic synthesis and for research applications. It is commonly used as a building block in the synthesis of substituted benzaldehydes, benzyl derivatives, and other nitroaromatic intermediates.

  • used as an intermediate in organic synthesis
  • chemical formula: C7H6BrNO2
  • molecular weight: 216.03 g/mol
  • cas number: 60956-26-5
  • purity typically around 98.7%
  • available in solid form, commonly supplied as a 100 g package

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.