Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-bromo-2-nitrotoluene | 60956-26-5 | MFCD00041243 | 98.7% | 216.03 g/mol | C7H6BrNO2 | 250 G
SDP

Catalog No. 5000208850
Encompass_Preferred

4-Bromo-2-nitrotoluene is an aromatic nitrotoluene reagent commonly used as a building block in organic synthesis and life-science research. The product is supplied as a solid powder with high reported purity and defined handling and storage recommendations.

  • High purity: 98.69%.
  • Molecular formula: C7H6BrNO2; molecular weight: 216.03 g/mol.
  • CAS number: 60956-26-5.
  • Appearance: light yellow to brown solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Pack size: 250 G.
  • Shipping: room temperature (may vary by region).

Catalog No. 50-002-08850 Supplier Medchemexpress LLC Supplier No. 250GHYW041357
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4-Bromo-2-nitrotoluene is an aromatic nitrotoluene reagent commonly used as a building block in organic synthesis and life-science research. The product is supplied as a solid powder with high reported purity and defined handling and storage recommendations.

  • High purity: 98.69%.
  • Molecular formula: C7H6BrNO2; molecular weight: 216.03 g/mol.
  • CAS number: 60956-26-5.
  • Appearance: light yellow to brown solid.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Pack size: 250 G.
  • Shipping: room temperature (may vary by region).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.