Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-bromo-2-nitrotoluene | 60956-26-5 | MFCD00041243 | 98.7% | 216.03 g/mol | C7H6BrNO2 | 500 G
SDP

Supplier:  Medchemexpress LLC HYW041357500G

Encompass_Preferred

4-Bromo-2-nitrotoluene is an organic reagent used as a building block in organic synthesis and life-science research. It is supplied as a solid of high purity for laboratory use and is labeled for research use only.

  • CAS number 60956-26-5.
  • Chemical formula C7H6BrNO2.
  • Molecular weight 216.03 g/mol.
  • Typical purity 98.7%.
  • Physical state solid.
  • Available in 500 G pack size.

Catalog No. 50-002-09554


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.