Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 4-Bromo-2-(trifluoromethyl)benzenol

Catalog No. KOB00231G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
KOB00231G 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. KOB00231G Supplier Sigma Aldrich Supplier No. KOB00231G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • The kinetics of gas-phase elimination of 1-(tert-butoxycarbonyl)-2-pyrrolidinone has been investigated in a static system
  • The observed rate coefficient has been reported to be log k1(s-1) = (11.54 +/- 0.29) -(140.8 +/- 2.8)-íkjmol-1 (2.303rt)-1

Specifications

CAS 50824-04-9
Molecular Formula C7H4BrF3O
Linear Formula C7H4BrF3O
MDL Number MFCD05668981
Quantity 25 mL
Molecular Weight (g/mol) 241.01
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.