Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-3,5-dichloropyridine, 95%, Thermo Scientific™

Catalog No. AAH6461606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6461606 5 g
AAH6461603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6461606 Supplier Thermo Scientific Chemicals Supplier No. H6461606

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 343781-45-3
Molecular Formula C5H2BrCl2N
Molecular Weight (g/mol) 226.88
MDL Number MFCD06797978
InChI Key LOKLQBZBVPYAQO-UHFFFAOYSA-N
Synonym 3,5-dichloro-4-bromo-pyridine, pubchem21381, 4-bromo-3,5-dichloropyridin, 3,5-dichloro-4-bromopyridine, pyridine,4-bromo-3,5-dichloro, 4-bromanyl-3,5-bis chloranyl pyridine
PubChem CID 10680788
IUPAC Name 4-bromo-3,5-dichloropyridine
SMILES ClC1=CN=CC(Cl)=C1Br

Specifications

Melting Point 75°C to 76°C
Color White
Quantity 5 g
UN Number UN2811
Beilstein 8760531
Formula Weight 226.88
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Bromo-3,5-dichloropyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.