Learn More
Selleck Chemical LLC 4-Bromo-3-hydroxybenzoic acid S5444-1g
SDP
Catalog No. 503108594
Encompass_Preferred
4-bromo-3-hydroxybenzoic acid is a potent inhibitor of the enzyme histidine decarboxylase with resultant inhibition of histamine formation in mammals
Catalog No. 50-310-8594 Supplier Selleck Chemical LLC Supplier No. S54441G
4-bromo-3-hydroxybenzoic acid is a potent inhibitor of the enzyme histidine decarboxylase with resultant inhibition of histamine formation in mammals