Learn More
Medchemexpress LLC 4-Bromo-3-hydroxybenzoic acid | 14348-38-0 | 217.018 | 1 G
SDP
Catalog No. 5000448721
Encompass_Preferred
4-Bromo-3-hydroxybenzoic acid is a metabolite of Brocresine and a histidine decarboxylase (HDC) inhibitor, with an IC50 of 1 mM for both rat fetal and rat gastric HDC. It also inhibits aromatic-L-amino acid decarboxylase from hog kidney and rat gastric mucosa in vitro. This product is intended for research use only.
  • Acts as a histidine decarboxylase inhibitor
  • Inhibits aromatic-L-amino acid decarboxylase
  • Suitable for in vitro research applications
  • Solid appearance, white to off-white color
Catalog No. 50-004-48721 Supplier Medchemexpress LLC Supplier No. HYW0034451G
4-Bromo-3-hydroxybenzoic acid is a metabolite of Brocresine and a histidine decarboxylase (HDC) inhibitor, with an IC50 of 1 mM for both rat fetal and rat gastric HDC. It also inhibits aromatic-L-amino acid decarboxylase from hog kidney and rat gastric mucosa in vitro. This product is intended for research use only.
  • Acts as a histidine decarboxylase inhibitor
  • Inhibits aromatic-L-amino acid decarboxylase
  • Suitable for in vitro research applications
  • Solid appearance, white to off-white color