Learn More
Medchemexpress LLC 4-Bromo-3-hydroxybenzoic acid | 14348-38-0 | 217.02 | 5 G
SDP
Catalog No. 5000449003
Encompass_Preferred
4-Bromo-3-hydroxybenzoic acid is identified as a metabolite of Brocresine. It acts as a histidine decarboxylase (HDC) inhibitor, demonstrating IC50s of 1 mM against both rat fetal and rat gastric HDC. Additionally, it inhibits aromatic-L-amino acid decarboxylase from hog kidney and rat gastric mucosa in vitro, with IC50s of 1 mM for both enzymes.
  • Metabolite of Brocresine
  • Inhibits rat fetal histidine decarboxylase (HDC) with an IC50 of 1 mM
  • Inhibits rat gastric HDC with an IC50 of 1 mM
  • Inhibits hog kidney aromatic-L-amino acid decarboxylase with an IC50 of 1 mM
  • Inhibits rat gastric mucosa aromatic-L-amino acid decarboxylase with an IC50 of 1 mM
Catalog No. 50-004-49003 Supplier Medchemexpress LLC Supplier No. HYW0034455G
Add to Cart
Add to Cart
4-Bromo-3-hydroxybenzoic acid is identified as a metabolite of Brocresine. It acts as a histidine decarboxylase (HDC) inhibitor, demonstrating IC50s of 1 mM against both rat fetal and rat gastric HDC. Additionally, it inhibits aromatic-L-amino acid decarboxylase from hog kidney and rat gastric mucosa in vitro, with IC50s of 1 mM for both enzymes.
  • Metabolite of Brocresine
  • Inhibits rat fetal histidine decarboxylase (HDC) with an IC50 of 1 mM
  • Inhibits rat gastric HDC with an IC50 of 1 mM
  • Inhibits hog kidney aromatic-L-amino acid decarboxylase with an IC50 of 1 mM
  • Inhibits rat gastric mucosa aromatic-L-amino acid decarboxylase with an IC50 of 1 mM