Learn More
Medchemexpress LLC 4-bromo-3-hydroxybenzoic acid | 14348-38-0 | 217.02 | 500 MG
SDP
Catalog No. 5000449144
Encompass_Preferred
4-bromo-3-hydroxybenzoic acid is a metabolite of Brocresine and acts as a histidine decarboxylase (HDC) inhibitor, with IC50s of 1 mM for both rat fetal and rat gastric HDC. It also inhibits aromatic-L-amino acid decarboxylase from hog kidney and rat gastric mucosa in vitro with IC50s of 1 mM for both enzymes.
  • Functions as a histidine decarboxylase (HDC) inhibitor
  • Also inhibits aromatic-L-amino acid decarboxylase
  • It is a metabolite of Brocresine
  • Available in various quantities
  • Exhibits high purity
  • Accompanied by documentation (data sheet, COA, SDS, handling instructions)
  • Soluble in DMSO
  • Intended for research use only
Catalog No. 50-004-49144 Supplier Medchemexpress LLC Supplier No. HYW003445500MG
4-bromo-3-hydroxybenzoic acid is a metabolite of Brocresine and acts as a histidine decarboxylase (HDC) inhibitor, with IC50s of 1 mM for both rat fetal and rat gastric HDC. It also inhibits aromatic-L-amino acid decarboxylase from hog kidney and rat gastric mucosa in vitro with IC50s of 1 mM for both enzymes.
  • Functions as a histidine decarboxylase (HDC) inhibitor
  • Also inhibits aromatic-L-amino acid decarboxylase
  • It is a metabolite of Brocresine
  • Available in various quantities
  • Exhibits high purity
  • Accompanied by documentation (data sheet, COA, SDS, handling instructions)
  • Soluble in DMSO
  • Intended for research use only