Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-3-iodotoluene 96.0+%, TCI America™
SDP

Catalog No. B51685G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51685G 5 g
B51681G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51685G Supplier TCI America Supplier No. B51685G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 858841-53-9
Molecular Formula C7H6BrI
Molecular Weight (g/mol) 296.93
MDL Number MFCD07784420
InChI Key FLOUOTRVXNQGKP-UHFFFAOYSA-N
PubChem CID 16218088
IUPAC Name 1-bromo-2-iodo-4-methylbenzene
SMILES CC1=CC(I)=C(Br)C=C1

Specifications

Boiling Point 150°C
Quantity 5 g
Formula Weight 296.93
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-3-iodotoluene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.