Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-3-methyl-5-(trifluoromethyl)pyrazole 98.0+%, TCI America™
SDP

Catalog No. B47701G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B47701G Supplier TCI America Supplier No. B47701G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 60061-68-9
Molecular Formula C5H4BrF3N2
Molecular Weight (g/mol) 229
MDL Number MFCD08060951
InChI Key PDSOUBXNWWZCNB-UHFFFAOYSA-N
Synonym 4-bromo-3-methyl-5-trifluoromethyl-1h-pyrazole, 4-bromo-5-methyl-3-trifluoromethyl-1h-pyrazole, 4-bromo-5-methyl-3-trifluoromethyl pyrazole, 4-bromo-5-methyl-3-trifluoromethylpyrazole, 1h-pyrazole, 4-bromo-3-methyl-5-trifluoromethyl, 4-bromo-3-trifluoromethyl-5-methyl pyrazole, 4-bromo-3-methyl-5-trifluoromethyl pyrazole, 4-bromo-3-methyl-5-trifluoromethyl-2h-pyrazole, acmc-20aiyv, 4-bromo-3-trifluoromethyl-5-methylpyrazole
PubChem CID 2736430
IUPAC Name 4-bromo-5-methyl-3-(trifluoromethyl)-1H-pyrazole
SMILES CC1=C(C(=NN1)C(F)(F)F)Br

Specifications

Melting Point 107°C
Color White-Yellow
Quantity 1 g
Formula Weight 229.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3-methyl-5-(trifluoromethyl)pyrazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.