Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-3-methylbenzoic acid, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
7697-28-1
C8H7BrO2
215.05
MFCD00039526
KWVXDZLVCISXIB-UHFFFAOYSA-N
4-bromo-m-toluic acid,4-bromo-3-methyl-benzoic acid,4-bromo-3-methylbenzoicacid,3-methyl-4-bromobenzoic acid,4-bromo-3-methyl benzoic acid,benzoic acid, 4-bromo-3-methyl,pubchem3807,4bromo-3-methylbenzoic acid,4-bromo-5-methylbenzoic acid,ksc379g0b
82131
4-bromo-3-methylbenzoic acid
CC1=C(C=CC(=C1)C(=O)O)Br
7697-28-1
C8H7BrO2
215.05
MFCD00039526
KWVXDZLVCISXIB-UHFFFAOYSA-N
4-bromo-m-toluic acid,4-bromo-3-methyl-benzoic acid,4-bromo-3-methylbenzoicacid,3-methyl-4-bromobenzoic acid,4-bromo-3-methyl benzoic acid,benzoic acid, 4-bromo-3-methyl,pubchem3807,4bromo-3-methylbenzoic acid,4-bromo-5-methylbenzoic acid,ksc379g0b
82131
4-bromo-3-methylbenzoic acid
CC1=C(C=CC(=C1)C(=O)O)Br
Color Tan to Yellow
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 5g
Melting Point 215.5°C to 216.4°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (HPLC)
Formula Weight 215.05
Physical Form Powder
Percent Purity 97%
Chemical Name or Material 4-Bromo-3-methylbenzoic acid
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 231-713-7