Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4'-Bromo-3'-nitroacetophenone 98.0+%, TCI America™
SDP

Catalog No. B33831G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B33831G 1 g
B33835G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B33831G Supplier TCI America Supplier No. B33831G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 18640-58-9
Molecular Formula C8H6BrNO3
Molecular Weight (g/mol) 244.04
MDL Number MFCD00016985
InChI Key YFVOFFKNHQTQQE-UHFFFAOYSA-N
Synonym 1-4-bromo-3-nitrophenyl ethanone, 4'-bromo-3'-nitroacetophenone, 1-4-bromo-3-nitrophenyl ethan-1-one, 4-bromo-3-nitroacetophenone, 4-bromo-3'-nitroacetophenone, ethanone, 1-4-bromo-3-nitrophenyl, acmc-209eny, 3-nitro-4-bromoacetophenone, 4-bromo-3-nitro acetophenone, 3'-nitro-4'-bromoacetophenone
PubChem CID 87737
IUPAC Name 1-(4-bromo-3-nitrophenyl)ethan-1-one
SMILES CC(=O)C1=CC=C(Br)C(=C1)[N+]([O-])=O

Specifications

Melting Point 119°C
Quantity 1 g
Formula Weight 244.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Bromo-3′-nitroacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.