Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-4'-(diphenylamino)biphenyl 93.0+%, TCI America™
SDP

Catalog No. B2844250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B2844250MG 250 mg
B28441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B2844250MG Supplier TCI America Supplier No. B2844250MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 202831-65-0
Molecular Formula C24H18BrN
Molecular Weight (g/mol) 400.32
MDL Number MFCD09038499
InChI Key NKCKVJVKWGWKRK-UHFFFAOYSA-N
Synonym (4′C-Bromobiphenyl-4-yl)diphenylamine
PubChem CID 11315550
IUPAC Name 4'-bromo-N,N-diphenyl-[1,1'-biphenyl]-4-amine
SMILES BrC1=CC=C(C=C1)C1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 129°C
Quantity 250 mg
Formula Weight 400.32
Percent Purity ≥93.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-4′-(diphenylamino)biphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.