Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (4-bromo-5-methyl-pyrazol-1-yl)-acetic acid

Catalog No. CDS014374 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS014374 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS014374 Supplier Sigma Aldrich Supplier No. CDS014374100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Perfluorooctanes can be used as a fluorous solvent for aerobic oxidation of alcohols to carbonyl compounds in fluorous biphasic systems
  • It can also be employed in the synthesis of perfluoroalkyl-substituted bis(oxazolines)

Specifications

Molecular Formula C6H7BrN2O2
Linear Formula C6H7BrN2O2
MDL Number MFCD03074362
Quantity 1 g
Molecular Weight (g/mol) 219.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.