Learn More
Cayman Chemical 4-bromo A23187, Purity: ≥95%, CAS: 76455-48-6, 10 mg
SDP
Catalog No. 502985164
Encompass_Preferred
A non-fluorescent analog of the highly selective calcium ionophore A23187; used in experiments where fluorescent probes are employed
Catalog No. 50-298-5164 Supplier Cayman Chemical Supplier No. 2043810 MG
A non-fluorescent analog of the highly selective calcium ionophore A23187; used in experiments where fluorescent probes are employed