Learn More
Medchemexpress LLC HY-N6694 1mg Medchemexpress, 4-​Bromo A23187 CAS:76455-48-6 Purity:>98%
SDP
Catalog No. 501970457
Encompass_Preferred
Medchemexpress, HY-N6694 1mg 4-​Bromo A23187 CAS:76455-48-6 4-Bromo A23187 is a halogenated analog of the highly selective calcium ionophore A-23187. 4-Bromo A23187,a calcium modulator, induces apoptosis in different cells, including HL-60 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0457 Supplier Medchemexpress LLC Supplier No. HYN66941MG
Medchemexpress, HY-N6694 1mg 4-​Bromo A23187 CAS:76455-48-6 4-Bromo A23187 is a halogenated analog of the highly selective calcium ionophore A-23187. 4-Bromo A23187,a calcium modulator, induces apoptosis in different cells, including HL-60 cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.