Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-m-xylene 97.0+%, TCI America™
SDP

Catalog No. B099625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B099625G 25 g
B0996250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B099625G Supplier TCI America Supplier No. B099625G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 583-70-0 | C8H9Br | 185.06 g/mol

Chemical Identifiers

CAS 583-70-0
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.06
MDL Number MFCD00000071
InChI Key YSFLQVNTBBUKEA-UHFFFAOYSA-N
Synonym 4-bromo-m-xylene, 2,4-dimethylbromobenzene, benzene, 1-bromo-2,4-dimethyl, 4-bromo-1,3-dimethylbenzene, 1,3-dimethyl-4-bromobenzene, m-xylene, 4-bromo, 1-bromo-2,4-dimethyl-benzene, 4-brom-m-xylol, pubchem3762, acmc-1aowv
PubChem CID 68503
IUPAC Name 1-bromo-2,4-dimethylbenzene
SMILES CC1=CC=C(Br)C(C)=C1

Specifications

Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
Boiling Point 208°C
Formula Weight 185.06
Percent Purity ≥97.0% (GC)
Grade Not Available
Chemical Name or Material 4-Bromo-m-xylene
EINECSNumber (3)-4042
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.