Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-N-[3-(trifluoromethyl)phenyl]benzamide, 97%, Thermo Scientific™

Catalog No. AAH58046ME
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH58046ME 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH58046ME Supplier Thermo Scientific Chemicals Supplier No. H58046ME

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Notes
Store in a dry, cool conditions in a well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56661-32-6
Molecular Formula C14H9BrF3NO
Molecular Weight (g/mol) 344.131
MDL Number MFCD00421306
InChI Key URMINAWTHPPDQD-UHFFFAOYSA-N
Synonym 4-bromo-n-3-trifluoromethyl phenyl benzamide, benzamide, 4-bromo-n-3-trifluoromethyl phenyl, 4-bromophenyl-n-3-trifluoromethyl phenyl carboxamide
PubChem CID 788568
IUPAC Name 4-bromo-N-[3-(trifluoromethyl)phenyl]benzamide
SMILES C1=CC(=CC(=C1)NC(=O)C2=CC=C(C=C2)Br)C(F)(F)F

Specifications

Quantity 500 mg
Formula Weight 344.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Bromo-N-[3-(trifluoromethyl)phenyl]benzamide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.