Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-N-Fmoc-L-phenylalanine, 95%

Catalog No. AAH5197806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5197806 5 g
AAH5197803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5197806 Supplier Thermo Scientific Chemicals Supplier No. H5197806
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 198561-04-5 | C24H20BrNO4 | 466.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 198561-04-5
Molecular Formula C24H20BrNO4
Molecular Weight (g/mol) 466.33
MDL Number MFCD00273460
InChI Key TVBAVBWXRDHONF-QFIPXVFZSA-N
Synonym fmoc-l-4-bromophenylalanine, s-n-fmoc-4-bromophenylalanine, fmoc-phe 4-br-oh, fmoc-4-bromo-l-phenylalanine, fmoc-l-4-br-phe-oh, fmoc-p-bromo-phe-oh, fmoc-d-4-bromophenylalanine, fmoc-l-4-bromophe, fmoc-l-phe 4-br-oh, fmoc-4'-bromo-l-phe
PubChem CID 2734449
SMILES OC(=O)[C@H](CC1=CC=C(Br)C=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 165°C to 168°C
Quantity 5 g
IUPAC Name (2S)-3-(4-bromophenyl)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)propanoic acid
Formula Weight 466.33
Percent Purity 95%
Chemical Name or Material 4-Bromo-N-Fmoc-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.