Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-N-methoxy-N-methylbenzamide 95.0+%, TCI America™
SDP

Catalog No. B57285G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B57285G 5 g
B57281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B57285G Supplier TCI America Supplier No. B57285G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 192436-83-2
Molecular Formula C9H10BrNO2
Molecular Weight (g/mol) 244.088
MDL Number MFCD02684303
InChI Key YEHZWOBPONLEEH-UHFFFAOYSA-N
PubChem CID 10999252
IUPAC Name 4-bromo-N-methoxy-N-methylbenzamide
SMILES CN(C(=O)C1=CC=C(C=C1)Br)OC

Specifications

Color Yellow
Quantity 5 g
Formula Weight 244.09
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-N-methoxy-N-methylbenzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.