Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-N,N,3,5-tetramethylaniline 98.0+%, TCI America™
SDP

Catalog No. B30705G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30705G 5 g
B30701G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30705G Supplier TCI America Supplier No. B30705G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 14275-09-3 | C10H14BrN | 228.13 g/mol

Chemical Identifiers

CAS 14275-09-3
Molecular Formula C10H14BrN
Molecular Weight (g/mol) 228.13
MDL Number MFCD11036245
InChI Key AQJHDBKXJXXDKV-UHFFFAOYSA-N
Synonym 2-Bromo-5-(dimethylamino)-m-xylene, 4-Bromo-N,N-dimethyl-3,5-xylidine
PubChem CID 44629944
IUPAC Name 4-bromo-N,N,3,5-tetramethylaniline
SMILES CN(C)C1=CC(C)=C(Br)C(C)=C1

Specifications

Color White-Yellow
Quantity 5 g
Packaging Glass Bottle
Physical Form Crystal-Powder at 20°C
Formula Weight 228.13
Percent Purity ≥98.0% (GC)
Grade Not Available
Chemical Name or Material 4-Bromo-N,N,3,5-tetramethylaniline
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.