Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromo-N,N-diethylaniline, 97%

Catalog No. AAL0361909
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0361909 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAL0361909 Supplier Thermo Scientific Chemicals Supplier No. L0361909
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2052-06-4 | C10H14BrN | 228.13 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Light sensitive. Store in the dark. Store away from oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2052-06-4
Molecular Formula C10H14BrN
Molecular Weight (g/mol) 228.13
MDL Number MFCD00013530
InChI Key NGYMZFJVHHKJQR-UHFFFAOYSA-N
PubChem CID 16328
IUPAC Name 4-bromo-N,N-diethylaniline
SMILES CCN(CC)C1=CC=C(C=C1)Br

Specifications

Melting Point 32°C to 35°C
Boiling Point 269°C to 271°C
Quantity 10 g
UN Number UN2811
Beilstein 2803424
Sensitivity Light sensitive
Synonym p-bromo-n,n-diethylaniline, benzenamine, 4-bromo-n,n-diethyl, n,n-diethyl-p-bromoaniline, 4-n,n-diethylamino-bromobenzene, benzenamine,4-bromo-n,n-diethyl, p-n,nddiethylaniline, 4-bromodiethylaniline, 4-bromophenyldiethylamine, 4-bromophenyl diethylamine, 4-bromo-phenyl diethylamine
Solubility Information Insoluble in water.
Formula Weight 228.13
Percent Purity 97%
Chemical Name or Material 4-Bromo-N,N-diethylaniline
Show More Show Less
Hazard Category H302+H332-H311
Hazard Statement GHS H Statement
H311-H302-H332
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber 218-140-8
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.