Learn More
AdipoGen 4-Bromo-naphthalic anhydride
SDP
Catalog No. 502561593
Encompass_Preferred
Chemical. CAS 81-86-7. Formula C12H5BrO3. MW 277.07. Synthetic. Building block for synthesis. Used for the preparation of fluorescent polymer compounds.
Catalog No. 50-256-1593 Supplier AdipoGen Supplier No. CDXB0425G005
Chemical. CAS 81-86-7. Formula C12H5BrO3. MW 277.07. Synthetic. Building block for synthesis. Used for the preparation of fluorescent polymer compounds.