Learn More
Medchemexpress LLC 4'-bromo-resveratrol | 1224713-90-9 | 99.9% | 291.14 g·mol^-1 | C14H11BrO2 | 10MG
SDP
Catalog No. 5000235817
Encompass_Preferred
4'-Bromo-resveratrol is a brominated derivative of resveratrol that functions as a potent dual inhibitor of sirtuin-1 (SIRT1) and sirtuin-3 (SIRT3). It is intended for research use in biochemical and cell-based assays probing sirtuin regulation, apoptosis, and metabolic reprogramming, and is supplied as a high-purity solid suitable for analytical and biological studies.
  • Potent dual inhibitor of SIRT1 and SIRT3.
  • High purity suitable for research applications.
  • Molecular formula C14H11BrO2; molecular weight 291.14 g·mol^-1.
  • Available in small quantities for assay development and screening.
  • Stable solid under recommended storage conditions.
Catalog No. 50-002-35817 Supplier Medchemexpress LLC Supplier No. HY12411310MG
4'-Bromo-resveratrol is a brominated derivative of resveratrol that functions as a potent dual inhibitor of sirtuin-1 (SIRT1) and sirtuin-3 (SIRT3). It is intended for research use in biochemical and cell-based assays probing sirtuin regulation, apoptosis, and metabolic reprogramming, and is supplied as a high-purity solid suitable for analytical and biological studies.
  • Potent dual inhibitor of SIRT1 and SIRT3.
  • High purity suitable for research applications.
  • Molecular formula C14H11BrO2; molecular weight 291.14 g·mol^-1.
  • Available in small quantities for assay development and screening.
  • Stable solid under recommended storage conditions.