Learn More
Medchemexpress LLC 4'-bromo-resveratrol | 1224713-90-9 | 99.9% | 291.14 | C14H11BrO2 | 50MG
SDP
Catalog No. 5000235088
Encompass_Preferred
4'-Bromo-resveratrol is a brominated derivative of resveratrol that functions as a potent, dual inhibitor of sirtuin-1 (SIRT1) and sirtuin-3 (SIRT3). It is used in biochemical and cellular research to study sirtuin-related epigenetic regulation and cancer pathways, including effects on melanoma cell growth, cell cycle, and apoptosis. The compound is supplied as a high-purity solid and is available in small research quantities and solution formats for assay use.
  • Potent dual inhibitor of sirtuin-1 and sirtuin-3.
  • Used to study mitochondrial metabolic reprogramming and antiproliferative effects in melanoma cells.
  • High purity solid with white to off-white appearance.
  • Available in small research quantities and solution formats suitable for assays.
  • Stable under recommended storage conditions; protect from light.
Catalog No. 50-002-35088 Supplier Medchemexpress LLC Supplier No. HY12411350MG
4'-Bromo-resveratrol is a brominated derivative of resveratrol that functions as a potent, dual inhibitor of sirtuin-1 (SIRT1) and sirtuin-3 (SIRT3). It is used in biochemical and cellular research to study sirtuin-related epigenetic regulation and cancer pathways, including effects on melanoma cell growth, cell cycle, and apoptosis. The compound is supplied as a high-purity solid and is available in small research quantities and solution formats for assay use.
  • Potent dual inhibitor of sirtuin-1 and sirtuin-3.
  • Used to study mitochondrial metabolic reprogramming and antiproliferative effects in melanoma cells.
  • High purity solid with white to off-white appearance.
  • Available in small research quantities and solution formats suitable for assays.
  • Stable under recommended storage conditions; protect from light.