Learn More
Medchemexpress LLC 4'-bromo-resveratrol | 1224713-90-9 | MFCD00238583 | 99.9% | 291.14 g/mol | C14H11BrO2 | 5MG
SDP
Catalog No. 5000235357
Encompass_Preferred
4'-Bromo-resveratrol is a brominated derivative of resveratrol used in research as a dual inhibitor of the sirtuin deacetylases SIRT1 and SIRT3. It is applied in biochemical and cellular studies to probe mitochondrial function, metabolic reprogramming, cell cycle regulation, and apoptosis signaling.
  • Dual inhibitor of SIRT1 and SIRT3 suitable for enzymatic and cellular assays.
  • High purity (~99.9%) for reliable analytical results.
  • Available as solid material and as DMSO solutions for flexible use.
  • Recommended storage at 4°C, protect from light; in solvent store at -80°C for long term.
  • Used to study mitochondrial function, cell proliferation, and apoptosis pathways.
Catalog No. 50-002-35357 Supplier Medchemexpress LLC Supplier No. HY1241135MG
4'-Bromo-resveratrol is a brominated derivative of resveratrol used in research as a dual inhibitor of the sirtuin deacetylases SIRT1 and SIRT3. It is applied in biochemical and cellular studies to probe mitochondrial function, metabolic reprogramming, cell cycle regulation, and apoptosis signaling.
  • Dual inhibitor of SIRT1 and SIRT3 suitable for enzymatic and cellular assays.
  • High purity (~99.9%) for reliable analytical results.
  • Available as solid material and as DMSO solutions for flexible use.
  • Recommended storage at 4°C, protect from light; in solvent store at -80°C for long term.
  • Used to study mitochondrial function, cell proliferation, and apoptosis pathways.