Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromobenzenesulfonyl Chloride 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
98-58-8
C6H4BrClO2S
255.51
MFCD00007437
KMMHZIBWCXYAAH-UHFFFAOYSA-N
4-bromobenzene-1-sulfonyl chloride,p-bromobenzenesulfonyl chloride,4-bromo-benzenesulfonyl chloride,p-bromophenylsulfonyl chloride,benzenesulfonyl chloride, 4-bromo,4-bromobenzenesulphonyl chloride,4-bromobenzenesulfonylchloride,unii-c6347t8fp1,4-bromobenzene sulfonyl chloride,4-bromophenyl chlorosulfone
7396
4-bromobenzenesulfonyl chloride
C1=CC(=CC=C1S(=O)(=O)Cl)Br
98-58-8
C6H4BrClO2S
255.51
MFCD00007437
KMMHZIBWCXYAAH-UHFFFAOYSA-N
4-bromobenzene-1-sulfonyl chloride,p-bromobenzenesulfonyl chloride,4-bromo-benzenesulfonyl chloride,p-bromophenylsulfonyl chloride,benzenesulfonyl chloride, 4-bromo,4-bromobenzenesulphonyl chloride,4-bromobenzenesulfonylchloride,unii-c6347t8fp1,4-bromobenzene sulfonyl chloride,4-bromophenyl chlorosulfone
7396
4-bromobenzenesulfonyl chloride
C1=CC(=CC=C1S(=O)(=O)Cl)Br
Boiling Point 153°C (15.0mmHg)
Quantity 25g
Infrared Spectrum Authentic
Color White to Beige
Melting Point 74°C to 77°C
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula BrC6H4SO2Cl
Beilstein 11, 57
Merck Index 15, 1414
Solubility Information Solubility in water: decomposes
Formula Weight 255.51
Physical Form Crystals or Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 4-Bromobenzenesulfonyl chloride, 98%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 202-683-2

TSCA : TSCA