Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4-bromobenzoic acid-d4 | 787624-24-2 | MFCD06658868 | 98.1% | 205.04 | C7HD4BrO2 | 250 MG
SDP

Catalog No. 5000280852
Encompass_Preferred

4-Bromobenzoic acid-d4 is the deuterium-labeled isotopologue of 4-bromobenzoic acid supplied for analytical and synthetic research. It is intended for use as a tracer, internal standard, or reagent in analytical methods and small-molecule synthesis.

  • Deuterium-labeled 4-bromobenzoic acid for research applications.
  • Suitable as a tracer or internal standard in analytical methods.
  • Available as powder and solution forms, multiple pack sizes including 250 mg.
  • High purity, 98.1%.
  • Molecular weight 205.04; formula C7HD4BrO2; CAS 787624-24-2.
  • Storage: powder at -20°C or 4°C; in solvent at -80°C or -20°C.
  • Intended for laboratory research use only, not for human use.

Catalog No. 50-002-80852 Supplier Medchemexpress LLC Supplier No. HYW249257250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

4-Bromobenzoic acid-d4 is the deuterium-labeled isotopologue of 4-bromobenzoic acid supplied for analytical and synthetic research. It is intended for use as a tracer, internal standard, or reagent in analytical methods and small-molecule synthesis.

  • Deuterium-labeled 4-bromobenzoic acid for research applications.
  • Suitable as a tracer or internal standard in analytical methods.
  • Available as powder and solution forms, multiple pack sizes including 250 mg.
  • High purity, 98.1%.
  • Molecular weight 205.04; formula C7HD4BrO2; CAS 787624-24-2.
  • Storage: powder at -20°C or 4°C; in solvent at -80°C or -20°C.
  • Intended for laboratory research use only, not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.