Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 4 -Bromoflavone 250mg | 20525-20-6 | 250 MG
SDP

Catalog No. 5000227959
Encompass_Preferred

4′-Bromoflavone (2-(4-bromophenyl)chromen-4-one) is a brominated flavone used in research as a chemopreventive agent and a potent inducer of phase II detoxification enzymes. It is supplied as a solid powder for laboratory use and is not intended for human or clinical applications.

  • Potent inducer of phase II detoxification enzymes and quinone reductase.
  • Reported inhibitor of CYP1A1 activity.
  • High purity (97.66%) suitable for biochemical and enzymatic assays.
  • Molecular formula C15H9BrO2; molecular weight 301.13 g·mol-1.
  • Supplied as a 250 mg solid for small-scale experiments.
  • Storage: powder -20°C for long-term; in solvent -80°C for extended storage.

Catalog No. 50-002-27959 Supplier Medchemexpress LLC Supplier No. HY115772250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

4′-Bromoflavone (2-(4-bromophenyl)chromen-4-one) is a brominated flavone used in research as a chemopreventive agent and a potent inducer of phase II detoxification enzymes. It is supplied as a solid powder for laboratory use and is not intended for human or clinical applications.

  • Potent inducer of phase II detoxification enzymes and quinone reductase.
  • Reported inhibitor of CYP1A1 activity.
  • High purity (97.66%) suitable for biochemical and enzymatic assays.
  • Molecular formula C15H9BrO2; molecular weight 301.13 g·mol-1.
  • Supplied as a 250 mg solid for small-scale experiments.
  • Storage: powder -20°C for long-term; in solvent -80°C for extended storage.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.