Learn More
Selleck Chemical LLC 4-Bromofluorobenzene-d4 E8262-5MG
SDP
Catalog No. 5000040903
Encompass_Preferred
4-Bromofluorobenzene-d4 is the deuterated product of 1-Bromo-4-fluorobenzene 1-Bromo-4-fluorobenzene is an organic halogenated aromatic compound mainly used in the synthesis of pharmaceutical intermediates or functional materials
Catalog No. 50-000-40903 Supplier Selleck Chemical LLC Supplier No. E82625MG
4-Bromofluorobenzene-d4 is the deuterated product of 1-Bromo-4-fluorobenzene 1-Bromo-4-fluorobenzene is an organic halogenated aromatic compound mainly used in the synthesis of pharmaceutical intermediates or functional materials