Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromofluorobenzene, MilliporeSigma™ Supelco™

Catalog No. 111020627 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1000 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-102-0627 1000 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 11-102-0627 Supplier MilliporeSigma Supelco Supplier No. 442404
This item is not returnable. View return policy

Sigma-Aldrich offers 4-bromofluorobenzene (1-Bromo-4-fluorobenzene) for a variety of applications that include, but are not limited to, environmental, food and beverage, forensics, petrochemical and pharmaceutical analyzes.

Environmental
TRUSTED_SUSTAINABILITY

Specifications

Packaging Ampule
CAS 460-00-4
Density 1.593 g/mL (at 25°C)
Flash Point 53°C
Physical Form Neat
Linear Formula BrC6H4 F
MDL Number MFCD00000342
Melting Point -16°C
Boiling Point 150°C
Synonym 1-Bromo-4-fluorobenzene
Type Organic Pollutant Standard
Quantity 1000 mg
Formula Weight 175.00
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.