Learn More
Medchemexpress LLC 4-bromoiodobenzene-d4 | 1147565-46-5 | 99.7% | 286.93 g/mol | C6D4BrI | 250 MG
SDP
Catalog No. 5000284714
Encompass_Preferred
  • Deuterium-labeled (d4) isotopologue for clear mass shift in MS and distinct NMR signals.
  • High chemical purity (≈99.7%) for reliable quantitative analysis.
  • Suitable as an internal standard or tracer in NMR, GC-MS, and LC-MS methods.
  • Stable under normal laboratory storage conditions when handled per safety guidance.
  • Available in multiple package sizes, including 250 mg, for flexible experimental needs.
Catalog No. 50-002-84714 Supplier Medchemexpress LLC Supplier No. HYW643348250MG
  • Deuterium-labeled (d4) isotopologue for clear mass shift in MS and distinct NMR signals.
  • High chemical purity (≈99.7%) for reliable quantitative analysis.
  • Suitable as an internal standard or tracer in NMR, GC-MS, and LC-MS methods.
  • Stable under normal laboratory storage conditions when handled per safety guidance.
  • Available in multiple package sizes, including 250 mg, for flexible experimental needs.