Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Bromomandelic acid, 98+%

Catalog No. AAA1297822
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1297822 100 g
AAA1297806 5 g
AAA1297814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1297822 Supplier Thermo Scientific Chemicals Supplier No. A1297822
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 6940-50-7 | C8H7BrO3 | 231.05 g/mol

4-Bromomandelic acid is used as a pharmaceutical intermediate. 4-bromomandelic acid is mainly used as an analytical reagent for zirconium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromomandelic acid is used as a pharmaceutical intermediate. 4-bromomandelic acid is mainly used as an analytical reagent for zirconium.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperatures and pressures.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6940-50-7
Molecular Formula C8H7BrO3
Molecular Weight (g/mol) 231.05
MDL Number MFCD00004232
InChI Key BHZBRPQOYFDTAB-UHFFFAOYNA-N
Synonym 4-bromomandelic acid, 2-4-bromophenyl-2-hydroxyacetic acid, p-bromomandelic acid, p-bromo mandelic acid, 4-bromo-dl-mandelic acid, benzeneacetic acid, 4-bromo-.alpha.-hydroxy, 4-bromophenyl hydroxy acetic acid, 4-bromo-alpha-hydroxyphenylacetic acid, 4-bromo-hydroxybenzeneaceticacid, mandelic acid, p-bromo
PubChem CID 97930
IUPAC Name 2-(4-bromophenyl)-2-hydroxyacetic acid
SMILES OC(C(O)=O)C1=CC=C(Br)C=C1

Specifications

Melting Point 115°C to 119°C
Quantity 100 g
Beilstein 1567909
Merck Index 14,1424
Solubility Information Slightly soluble in water.
Formula Weight 231.05
Percent Purity ≥98%
Chemical Name or Material 4-Bromomandelic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 230-085-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.